ProPette™ LE Adjustable Pipettors, MTC Bio
Supplier: MTC BIO, INC.
P5200-100U P5200-10M P5200-10U P5200-1M P5200-200U P5200-20U P5200-2U P5200-300U P5200-50U P5200-5M
76337-016EA 642.67 CAD
76337-016 76337-018 76337-116 76337-118 76337-120 76337-122 76337-124 76337-126 76337-128 76337-130
ProPette™ LE Adjustable Pipettors, MTC Bio
Pipettes Repetitive Pipettors
The ProPette LE™ is the next generation of high precision liquid handling instruments from MTC Bio. This addition to the Propette family features improved actuation forces and a nearly 20% reduction in overall weight.
  • Adjustable with Roto-Lock™
  • Light weight with reduced forces
  • Full volume range (0.1 µl to 10 ml)
  • Superior accuracy and reproducibility
  • Outstanding user comfort
Every aspect of accuracy, reliability and user comfort has been carefully reviewed in designing these high precision pipettes.  The result is an advanced pipette that exhibitsMore Product Information