One-Touch Pumps, Menda
Supplier: Desco
35301 35303
15431-760EA 42.27 CAD
15431-760 15431-764
One-Touch Pumps, Menda
Bottle Top Dispensers
The one-touch pump features precision machined valves for smooth and consistent operation.
  • Neck size: 38-400 thread
  • Patented one-handed operation frees operator's hands
  • Precision machined valves for smooth and consistent operation
  • Cam-lock hinge keeps lid open or closed, to reduce odors
  • Dispenses approximately 0.2 cc of liquid with each pump
  • Ideal for use with hand sanitizer
One-touch pumps are not recommended for use with acrylic liquid. No silicone or latex is used, making it perfect for ultra clean environments. Ideal for cleaning with cotton balls, pads, and brushes. Fits all standard Menda HDPE, glass, and steel bottles. 100% Stainless steel construction will not corrode or contaminate liquid.