Black Nylon Probes, Menda
Supplier: Desco
35619 35621 35622
15431-800EA 6.68 CAD
15431-800 15431-802 15431-804
Black Nylon Probes, Menda
Tool Sets
Black nylon probes are ideal for use in an ESD protected area.
  • Nylon finish will not mark delicate surfaces
  • Temperature resistant
Ideal for opening plastic cases such as MP3 players, cell phones, laptops, and more. Also a great tool for popping out batteries. May be suitable for use in ESD protected areas.
The hygroscopic (readily accepts moisture) properties of nylon may make this tool suitable for use around ESD sensitive components after a few minutes of handling the tool with bare hands. Without exposure to moisture or antistatic treatment,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable