Peptide Synthesis Vessel,s Solid Phase, With Side Port, Chemglass
Supplier: Chemglass
CG-1861-04 CG-1861-10 CG-1861-12 CG-1861-13 CG-1861-02 CG-1861-03 CG-1861-01 CG-1861-14 CG-1861-05 CG-1861-11 CG-1861-15 CG-1861-06
89029-990EA 763.1 CAD
89029-990 89029-992 89029-994 89029-996 89029-986 89029-988 10170-982 10170-986 10170-990 10170-992 10170-996 10171-002
Peptide Synthesis Vessel,s Solid Phase, With Side Port, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Solid phase peptide synthesis vessel having a medium or coarse porosity fritted glass resin support.
Top of the vessel has a GL thread and is supplied complete with a PTFE lined PBT screw cap. Side port and lower PTFE stopcock have a 2 mm bore with the arms being 8 mm I.D.
Ordering information: Vessels are available in sizes other than those listed.