Take-Along Pumps, Menda
Supplier: Desco
35700 35713 35714
15431-776EA 50.81 CAD
15431-776 15431-778 15431-780
Take-Along Pumps, Menda
Bottle Top Dispensers
The Menda stainless steel take-along pump features precision machined valves for smooth operation, as well as a locking pump that prevents leaks when engaged.
  • Neck size: 38-400 thread
  • Patented one handed operation frees operator's hands
  • Precision machined valves for smooth and consistent operation
  • Cam-lock hinge keeps lid open or closed, to reduce odors
  • Dispenses approximately 0.2 cc of liquid with each pump
Locking pump will not leak when engaged, and is suitable for travel. No silicone or latex is used, making it perfect for ultra clean environments. Ideal for cleaning with cotton balls, pads, and brushes. Fits all standard Menda HDPE, glass, and steel bottles. Genuine made in the United States of America Menda pump.