Pure-Take Pumps, Menda
Supplier: Desco
35800 35813
15431-784EA 53.28 CAD
15431-784 15431-786
Pure-Take Pumps, Menda
Bottle Top Dispensers
The pure-take pump features a purity protector to stop fluid from re-entering the bottle, as well as a locking pump to prevent leaks when engaged.
  • Neck size: 38-400 thread
  • Precision machined valves for smooth and consistent operation
  • Cam-lock hinge keeps lid open or closed, to reduce odors
  • Dispenses approximately 0.2 cc of liquid with each pump
Locking pump will not leak when engaged, making the device suitable for travel. No silicone or latex is used, making them perfect for ultra clean environments. Ideal for cleaning with cotton balls, pads, andMore Product Information