Wright-Giemsa stain solution APHA, HARLECO®, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms: Wright-Giemsa Solution
742-86 742-75 742-86 742-75
CA15204-232CS 2270.67 CAD
CA15204-232 15204-226 CA15204-226 15204-232
Wright-Giemsa stain solution APHA, HARLECO®, Sigma-Aldrich®
Wright-Giemsa stain (solutions & kits)
HARLECO® Polychromic stain for peripheral blood and bone marrow smears. Specifically formulated for staining malarial parasites.
  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use
Polychromic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation. Specifically formulated for staining malarial parasites.
IVDLogo