Wright-Giemsa stain solution Fucillo, HARLECO®, Sigma-Aldrich®
Supplier: MilliporeSigma
Synonyms: Wright-Giemsa Solution
64571-95 64571-75 64571-85 64571-95 64571-85 64571-75
CA15204-158EA 336.81 CAD
15204-158 CA15204-158 15204-159 CA15204-159 34172-051 CA34172-051
Wright-Giemsa stain solution Fucillo, HARLECO®, Sigma-Aldrich®
Wright-Giemsa stain (solutions & kits)
HARLECO® Polychromatic stain for peripheral blood and bone marrow smears
  • Prepared from dyes certified by the Biological Stain Commission (BSC)
  • For In-Vitro Diagnostic Use
  • For Professional Use
Polychromatic stain for peripheral blood and bone marrow smears. Used for red blood cell morphology and white blood cell differentiation.
IVDLogo