PTFE Joint Clips, Ace Glass Incorporated
Supplier: Ace Glass
7596-24 7596-29 7596-45
76336-706EA 66.57 CAD
76336-706 76336-708 76336-710
PTFE Joint Clips, Ace Glass Incorporated
Clamps Standard Taper and Spherical Joint Clamps
Keck® like, mechanical PTFE joint clips snap on and off with ease.
  • Do not scratch the glass and can be used to 250 °C
  • Can also be used for clamping stopcock plugs and barrels
  • ical PTFE joint clips snap on and off with ease
  • For slight pressure use only; below 5 psig
Manufactured from 3M™ Dyneon™ TF1620.
Certifications: These clips conform with FDA 21 CFR 177.1550 and USP Class VI.
Order Now
SPECIFICATIONS
Environmentally Preferable