Adapters, Distilling/Splitting, Process Reactors, Angled, Chemglass
Supplier: Chemglass
CG-1235-B-24 CG-1235-B-29
30622-530EA 1235.99 CAD
30622-530 30622-532
Adapters, Distilling/Splitting, Process Reactors, Angled, Chemglass
Distillation Components Distillation Adapters
Compact and simple distilling/splitter adapter for use with process reactors lids that have a 10° angled side neck.
  • Control valve is either a 0 to 4 mm or a 0 to 12 mm PTFE with a 1/2" OD distillate takeoff
  • For use with compression fittings and TFE tubing
  • Plugs are supplied complete with perfluoro O-rings
Vapor flows upward freely through the large vapors holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor.