Winged Infusion Sets, SAI Infusion Technologies
Supplier: SAI Infusion Technologies
WHIS-20-75
89134-232PK 290.4 CAD
89134-232
Winged Infusion Sets, SAI Infusion Technologies
Animal Supplies Animal Infusion Supplies
These winged infusion sets are designed for easy insertion into subcutaneous Vascular Access Ports (VAPS), and can be used for continuous or bolus infusions.
  • Huber needle allows easy penetration without coring
  • Alternate Huber needle configurations are available
  • Connect to standard syringes or a luered fitting
  • Customizable
Each set is comprised of a 90° Huber Needle with suture wings, connected to 10" of mirco-bore tubing, with a luer lock fitting. Supplied sterile.
Packaging: Packaged individually.