Flasks, Volumetric, Class A, Mixing Bulbs, Chemglass
Supplier: Chemglass
CG-1601-01 CG-1601-02 CG-1601-03 CG-1601-05
80070-366EA 266.41 CAD
80070-366 80070-368 80070-370 80070-374
Flasks, Volumetric, Class A, Mixing Bulbs, Chemglass
Flasks Volumetric Flasks
Similar to CG-1600 with the addition of a mixing bulb between the calibration line and the stopper.
  • The mixing bulb contains approximately one tenth of the flask capacity
  • The mixing bulbs aids in the mixing or dissolving of the flask contents
For additional stoppers, see CG-3018.
Ordering information: Supplied complete with one solid glass pennyhead stopper. For additional stoppers, see CG-3018.