Distilling Heads, Short Path, Chemglass
Supplier: Chemglass
CG-1239-04 CG-1239-13
80067-570EA 620.37 CAD
80067-570 80067-576 80067-580 80067-586
Distilling Heads, Short Path, Chemglass
Distillation Components Distillation Heads
Distilling heads are available with vigreux indentations.
  • The thermometer bulb and stem serve as column packing.
  • Upper hose connections are for cooling lines
  • The lower connection is for vacuum
The 14/20 size accepts a 1" immersion 7/12 or 10/18 thermometer, while the 24/40 and 29/42 sizes accepts a 2" immersion, 10/30 thermometer.