Cylinders, Graduated, with Outer Joints, Chemglass
Supplier: Chemglass
CG-1223-01 CG-1223-03 CG-1223-05 CG-1223-07 CG-1223-09 CG-1223-02 CG-1223-04
80067-450EA 281.53 CAD
80067-450 80067-454 80067-458 80067-462 80067-466 80067-452 80067-456
Cylinders, Graduated, with Outer Joints, Chemglass
Cylinders Measuring Cylinders
Cylinders are calibrated to contain at 20 °C.
  • Supplied without stopper
  • Tolerances and subdivisions are the same as CG-8242
Ordering information: For standard taper stoppers see CG-3000.