Funnels, Powder, Outer Joints, Chemglass
Supplier: Chemglass
CG-1762-02 CG-1762-04
89028-762EA 121.69 CAD
89028-762 89028-764
Funnels, Powder, Outer Joints, Chemglass
Funnels Powder Funnels
Funnels feature outer joints.
  • Funnels have 60° side angle
  • Standard taper outer joint
Powder funnel is useful for the introduction of solids or liquids into reactions under atmospheric conditions.