Distilling Receivers, Moisture Test, Merlic, Chemglass
Supplier: Chemglass
CG-1264-04 CG-1264-06
80067-826EA 789.15 CAD
80067-826 80067-830
Distilling Receivers, Moisture Test, Merlic, Chemglass
Distillation Components Distillation Receivers
Distillation receivers feature efficient cooling.
  • Graduated from 0 to 10 ml in 0.2 ml subdivisions
Modified Dean-Stark trap allows for more efficient cooling and separation of the azeotrope during distillation.
Order Now
SPECIFICATIONS
Environmentally Preferable