Flasks, Heavy Wall, Round Bottom, With Thermowells, Chemglass
Supplier: Chemglass
CG-1515-01 CG-1515-02 CG-1515-03
80069-090EA 504.07 CAD
80069-090 80069-092 80069-094
Flasks, Heavy Wall, Round Bottom, With Thermowells, Chemglass
Flasks Round Bottom Flasks
Heavy wall flasks feature a standard taper.
  • High-quality, round bottom
  • A side well to accept a thermometer
Top of thermowell is tooled to accept a #0 rubber stopper.