Condensers, 'C' Assembly, Constant Feed, for Rotary Evaporators, Plastic Coated, Chemglass
Supplier: Chemglass
CG-1333-A-02 CG-1333-A-01 CG-1333-A-15
47733-790EA 2635.5 CAD
47733-790 80060-964 80060-966
Condensers, 'C' Assembly, Constant Feed, for Rotary Evaporators, Plastic Coated, Chemglass
Condensers
Two piece cold trap style Condenser for use with rotary evaporators.
  • Condenser is supplied complete with all fittings necessary to connect the continuous feed feature
  • Along with Chemglass new style PTFE centering ring which makes a vacuum tight seal between the inner cold finger and the plastic coated outer sleeve
  • Also supplied with a UHMW lid with knob
Condenser has an additional 19/38 outer joint sealed onto the condenser to allow continuous feed of volumes exceeding the capacity of the evaporating flask. The 19/38 joint is sealed at an angle whichMore Product Information