Distillation Receivers, Chemglass
Supplier: Chemglass
CG-1281-01 CG-1281-14
80067-932EA 1891.56 CAD
80067-932 80067-942
Distillation Receivers, Chemglass
Distillation Components Distillation Receivers
These distillation receivers are similar to CG-1279 but with the top outer joint vertical instead of a 105º angle.
  • Complete item consists of a vacuum adapter, distribution adapter
Delivery information: Supplied with four of the listed size flasks with hooks and ten CG-110 stainless steel springs.