Reaction Vessels, Cylindrical, Jacketed, Chemglass
Supplier: Chemglass
CG-1926-01 CG-1926-02 CG-1926-03 CG-1926-04 CG-1926-05 CG-1926-06
80071-480EA 2174.68 CAD
80071-480 80071-482 80071-484 80071-486 80071-488 80071-490
Reaction Vessels, Cylindrical, Jacketed, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange.
  • Cooling jacket is equipped with two removable hose connections
  • Reduce the risk of injury while attaching or detaching flexible tubing
  • Supplied complete with two polypropylene hose connections and two screw caps of the listed size
Flange is also ground to permit a greased seal with any of the lids. clamp and O-ring is also available.
Ordering information: High temperature hose connections and screw caps are available. See CG-195-H AND CG-198-H.