Filter Apparatus, Chemglass
Supplier: Chemglass
CG-1404-01 CG-1404-02 CG-1404-03
80068-332EA 428.36 CAD
80068-332 80068-334 80068-336
Filter Apparatus, Chemglass
Funnels Filtration Funnels Filter Funnels
Useful for direct filtration applications of small volumes under vacuum.
  • Complete apparatus consists of a Buchner funnel
  • Featuring 14/20 top outer and lower inner joints
Erlenmeyer flask 50 ml having a 14/20 outer joint.