Inlet Feeds, Chemglass
Supplier: Chemglass
CG-1327-20 CG-1327-30
80060-930EA 278.88 CAD
80060-930 80068-138
Inlet Feeds, Chemglass
Adapters Adapters, Joints with Tube Connectors
Inlet feed tubes feature a bore stopcock PTFE.
  • Inlet feed tube for 'A' assemblies
  • Joint size is 19/38
Inlet feed feature a 90° bore stopcock and PTFE extension tube.