Filter Funnels, Hirsch, Chemglass
Supplier: Chemglass
CG-1408-06 CG-1408-02 CG-1408-03 CG-1408-04 CG-1408-07 CG-1408-08
80068-562EA 254.14 CAD
80068-562 80068-554 80068-556 80068-558 80068-564 80068-566
Filter Funnels, Hirsch, Chemglass
Funnels Filtration Funnels Filter Funnels
The 60º angle on the funnel permits removal of precipitates.
  • High-quality filter funnels, hirsch permits removal of precipitates
  • Wash and re-dissolve the precipitate with chemicals
Used in applications where it is necessary to wash and re-dissolve the precipitate with chemicals that would attack standard filter paper.