Distilling Receivers, Moisture Test, Overflow Tube, Dean-Stark, Chemglass
Supplier: Chemglass
CG-1259-01 CG-1259-03 CG-1259-04
80067-746EA 591.82 CAD
80067-746 80067-750 80067-752
Distilling Receivers, Moisture Test, Overflow Tube, Dean-Stark, Chemglass
Distillation Components Distillation Receivers
Distilling receivers feature a overflow tube.
  • Overflow tube
  • Between the uptake tube and the graduated receiver
Similar to CG-1258 but with the addition of an overflow tube between the uptake tube and the graduated receiver.