Distilling Receivers, Moisture Test, Barrett, Overflow Tube, Chemglass
Supplier: Chemglass
CG-1263-05 CG-1263-06 CG-1263-10 CG-1263-11
80067-812EA 829.01 CAD
80067-812 80067-814 80067-816 80067-818
Distilling Receivers, Moisture Test, Barrett, Overflow Tube, Chemglass
Distillation Components Distillation Receivers
Distillation receivers feature an overflow tube.
  • Overflow tube between the uptake tube and the graduated receiver
Similar to Barrett type receivers but with the addition of an overflow tube between the uptake tube and the graduated receiver.