Adapters, Distilling/Splitting, Process Reactors, Chemglass
Supplier: Chemglass
CG-1235-A-24 CG-1235-A-29
30622-524EA 1218.01 CAD
30622-524 30622-526
Adapters, Distilling/Splitting, Process Reactors, Chemglass
Distillation Components Distillation Adapters
Compact and simple distilling/splitter adapter for use with process reactors.
  • Control valve is 0 to 4 mm PTFE
  • A 1/2" O.Ø distillate takeoff for use with compression fittings and TFE tubing
  • Plugs are supplied complete with Perfluoro O-rings
Vapor flows upward freely through the large vapor holes to the condenser. Distillate flow is directed using the valve to either the receiving flask or back to the reactor.