Funnels, Heavy Duty, Chemglass
Supplier: Chemglass
CG-1768-02 CG-1768-05 CG-1768-01 CG-1768-03 CG-1768-04
89028-784EA 243.13 CAD
89028-784 89028-790 89028-792 89028-782 89028-786 89028-788
Funnels, Heavy Duty, Chemglass
Funnels Filtration Funnels Analytical Funnels
Heavy duty funnels are autoclavable.
  • Highly recommended for continuous laboratory work
  • Steam autoclavable at 121 °C (250 °F)
Sturdy polypropylene funnels with ridges to prevent vapor lock.