Flasks, Heavy Wall, Round Bottom, 4-Necks, With Threaded Side Arm, Chemglass
Supplier: Chemglass
CG-1534-02 CG-1534-03 CG-1534-04 CG-1534-06 CG-1534-07 CG-1534-09 CG-1534-10 CG-1534-12 CG-1534-01 CG-1534-24
80069-944EA 685.56 CAD
80069-944 80069-946 80069-948 80069-952 80069-954 80069-958 80069-960 80069-964 80069-942 80069-966
Flasks, Heavy Wall, Round Bottom, 4-Necks, With Threaded Side Arm, Chemglass
Flasks Round Bottom Flasks
The round bottom, heavy wall, four-neck flask feature a standard taper.
  • For insertion of thermometers
  • For use with bleed tubes
  • Has an O.D. between 4 and 7 mm
Standard taper outer joints and a #7 Chem-Thread.