Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joints, Chemglass
Supplier: Chemglass
CG-1508-07 CG-1508-08 CG-1508-10 CG-1508-12 CG-1508-15 CG-1508-18 CG-1508-33 CG-1508-35 CG-1508-36 CG-1508-03 CG-1508-04 CG-1508-05 CG-1508-06 CG-1508-34
80068-906EA 210.21 CAD
80068-906 80068-908 80068-910 80068-912 80068-914 80068-916 80068-918 80068-920 80068-924 80068-926 80068-898 80068-900 80068-902 80068-904 80068-922
Flasks, Heavy Wall, Round Bottom, Single Neck, Spherical Joints, Chemglass
Flasks Round Bottom Flasks
Round bottom, heavy wall, single short neck flasks with a spherical socket joint.
  • Replacement flasks for the collection flasks
  • For use with replacement flasks for rotary evaporator collections
Flasks having a 35/20 socket joint are used as replacements for the collection flask on rotary evaporators.