Hematoxylin solution Gill II, Shandon™

Supplier: RICHARD-ALLAN SCIENTIFIC CA

Synonyms: DcH-48, Natural Black I, Hydroxybrasilin

6765007 6765008
CA89369-336CS 408.38 CAD
CA89369-336 CA89369-338
Hematoxylin solution Gill II, Shandon™
Hematoxylin staining solutions

Achieve fast and convenient results, while maintaining complete control over stain intensity with Thermo Scientific™ Shandon™ Gill™ Hematoxylin, which has mucoid staining.


  • Gill 1: Light blue nuclei
  • Gill 2: Medium blue nuclei
  • Gill 3: Blue-black nuclei


For darker nuclei or shorter staining times Gill 2 may be used

Formula: C₁₆H₁₄O₆
MW: 302.28 g/mol
MDL Number: MFCD00078111
CAS Number: 517-28-2
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR