Eosin Y (yellowish) alcoholic solution, Shandon™ cytoplasmic counterstain

Supplier: RICHARD-ALLAN SCIENTIFIC CA

Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

6766008 6766007 6766008
CA89212-376EA 503.29 CAD
89370-828 89212-376 CA89212-376
Eosin Y (yellowish) alcoholic solution, Shandon™ cytoplasmic counterstain
Eosin Y (yellowish)

The Epredia™ Shandon™ Eosin-Y Staining Solutions are fast-acting cytoplasmic counterstain.


  • Stable and precipitate-free; no filtering necessary
  • Produces consistent results
  • pH-adjusted for maximum stain uptake
  • Produces brilliant staining
  • Compatible with all alum hematoxylin stains
  • Available in alcoholic and aqueous formulations
  • Ready-to-use

Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
MDL Number: MFCD00005040
CAS Number: 17372-87-1
IVDLogo

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR