Papanicolaou's solution EA 65
Supplier: RICHARD-ALLAN SCIENTIFIC CA
Synonyms: EA 36
75604 75611 75625
CA84000-124CS 608.88 CAD
CA84000-124 CA84000-126 CA84000-128
Papanicolaou's solution EA 65
Papanicolaou's solution EA 65
Control the color hues and intensities of your work with Epredia™ Papanicolaou EA Stain, useful for both gynecological and non-gynecological staining.
  • EA-36 and EA-50 are used in conjunction with OG-6 for gynecological staining.
  • EA-65 is used with OG-6 for non-gynecological staining.
  • Modified EA is a Gill modification that minimizes precipitation, reduces staining time, and does not contain Bismarck Brown.
Epredia™ EA 36 is used in conjunction with Epredia™ OG 6 for the staining of gynecological specimens. It will provide a slightly lighter cytoplasmic hue than EA 50. EA 36 is recommended for end users who desire a more pastel staining result in gynecological specimens.
IVDLogo