Glas-Col® STM Series Mantles, High Temperature, Chemglass
Supplier: Chemglass
CG-12500-01 CG-12500-04 CG-12500-06 CG-12500-07 CG-12500-08 CG-12500-09
80060-732EA 1119.58 CAD
80060-732 80060-734 80060-736 80060-738 80060-740 80060-742
Glas-Col® STM Series Mantles, High Temperature, Chemglass
Heaters Heating Mantles
The Glas-Col® Series STM mantle performs the same function as a Series TM heating mantle, plus operations requiring more heat.
  • The STM mantle has a durable aluminum exterior and a resilient and pliable interior to cradle glassware
  • Withstanding an upper internal operating temperature of 650º C, the STM mantle is very resistant to burnout
  • A splash guard and poncho safety shield can be used to prevent damage from accidental spills
The higher temperature range enables operations to be performed that require more heat, such as rapid distillation of waterMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable