Reaction Vessels, Cylindrical, Jacketed, Lower Drain Valves, Chemglass
Supplier: Chemglass
CG-1930-23 CG-1930-24 CG-1930-25 CG-1930-26 CG-1930-27 CG-1930-20 CG-1930-22
47733-820EA 3159.12 CAD
47733-820 47733-822 47733-824 47733-826 47733-828 80071-516 80071-518
Reaction Vessels, Cylindrical, Jacketed, Lower Drain Valves, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Vessels feature a cylindrical jacketed lower drain valves.
  • Similar to CG-1924, but with a choice of CG-911-A 0 to14 mm sidearm valve
  • A straight through 0 to 12 mm CG-510 valve or 0 to 20 mm CG-511 sidearm valve
All valves are designed to seal flush with the bottom of the flask avoiding the dead space normally associated with stopcock drains. This feature permits total mixing and draining of the contents of the reactor. The CG-510 is recommended for solution based reactions while the CG-911-A and CG-511 are recommended for reactions containing anyMore Product Information