Wright's stain

Supplier: RICHARD-ALLAN SCIENTIFIC CA

89010 89012
CA84000-176EA 422.51 CAD
CA84000-176 CA84000-180
Wright's stain
Wright's stain

Utilize Epredia™ Wright Stain for peripheral blood and bone marrow smears.

MDL Number: MFCD00082143
CAS Number: 68988-92-1
IVDLogo

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR