ChemSplash® 1 Chemical Splash Coveralls with Hood and Boot, Elastic Wrist and Ankle, Taped Seams, Elastic Back, International Enviroguard™
Supplier: International Enviroguard
7019YT-M 7019YT-L 7019YT-XL 7019YT-2XL 7019T-3XL 7019YT-4XL
94001-188CS 318.46 CAD
94001-188 94001-190 94001-192 94001-194 94001-196 94001-198
ChemSplash® 1 Chemical Splash Coveralls with Hood and Boot, Elastic Wrist and Ankle, Taped Seams, Elastic Back, International Enviroguard™
Disposable Garments
Disposable Coverall designed to protect against chemical splash and toxic particles.
  • Proprietary hybrid membrane fabric helps to keep wearer cool and avoid heat stress
  • Taped seams improve protection vs coveralls made with serged seams
  • Contrasting red stripe to quickly distinguish this taped-seam garment from serged seam coveralls
Roomy, seamless shoulder area resists tears and provides more protection in key exposure area. Wider, gusseted crotch gives wearer more room to move in all directions without ripping fabric. Elastic back stretches with wearer for more comfort and better tear resistance. Storm flap over zipper with pressure sensitive tape adds an extra layer of protection. High moisture vapor transfer rate improves wearer comfort.
Recommended applications include manufacturing, emergency response, environmental clean-up, and more.