Funnels, Addition, Graduated, PTFE Metering Stopcocks, Chemglass
Supplier: Chemglass
CG-1713-05 CG-1713-11 CG-1713-16 CG-1713-19 CG-1713-20 CG-1713-21 CG-1713-01 CG-1713-03 CG-1713-04 CG-1713-06 CG-1713-07 CG-1713-09 CG-1713-10 CG-1713-12 CG-1713-13 CG-1713-14 CG-1713-15 CG-1713-17
80070-690EA 723.09 CAD
80070-690 80070-702 80070-712 80070-718 80070-720 80070-722 80070-682 80070-686 80070-688 80070-692 80070-694 80070-698 80070-700 80070-704 80070-706 80070-708 80070-710 80070-714
Funnels, Addition, Graduated, PTFE Metering Stopcocks, Chemglass
Funnels Dropping Funnels
Cylindrical style addition funnels feature a PTFE adjustable bore metering plug.
  • Turning the adjustment knob allows flow rate to be controlled from dropwise to full flow
For replacement stoppers see CG-3000.