Condensers, High Efficiency, Chemglass
Supplier: Chemglass
CG-1215-C-10 CG-1215-C-01 CG-1215-C-02
16002-676EA 4241.73 CAD
16002-676 80067-344 80067-346
Condensers, High Efficiency, Chemglass
Condensers
This unique condenser design combines features for refluxing in a single condenser.
  • Coolant, introduced into the upper hose connection
  • Fills a large cold finger that is wrapped with coils
  • Hose connections for the 24/40 size condenser are 10 mm at the largest serration
  • Hose connections for the 45/50 size are 15 mm at the largest serration
After the coolant travels through the coils, it enters the water jacket to maximize cooling surface along the vapor path. Angled take off joint is a standard taper 24/40, and the angled take off joint for the 45/50 size is a standard taper 29/42.