Micro Volumetric Flasks with Stopper, Class A, Chemglass
Supplier: Chemglass
CG-1603-01 CG-1603-02 CG-1603-03 CG-1603-04 CG-1603-05
80070-406EA 207.39 CAD
80070-406 80070-408 80070-410 80070-412 80070-414
Micro Volumetric Flasks with Stopper, Class A, Chemglass
Flasks Volumetric Flasks
Class 'A' micro volumetric flasks are supplied complete with CG-3000 14/20 hollow stopper.
  • Flasks have hex base for stability
  • Flasks have a 'V' shaped interior bottom for total sample retrieval
  • Calibrated to contain 14/20 top outer joint