Funnels, Separatory, Graduated, PTFE Stopcocks, Chemglass
Supplier: Chemglass
CG-1734-01 CG-1734-04 CG-1734-12 CG-1734-14 CG-1734-02 CG-1734-03 CG-1734-10
80071-012EA 608.98 CAD
80071-012 80071-018 80071-026 80071-028 80071-014 80071-016 80071-024
Funnels, Separatory, Graduated, PTFE Stopcocks, Chemglass
Funnels Separating Funnels
Graduated funnels feature a PTFE stopcock.
  • Cylindrical shaped separatory funnel having a lower standard taper inner joint
  • A top outer joint, both being the same size
Graduated with a double scale of numbers, one up from the stopcock and one down from the capacity line. Funnels with 14/20 and 19/22 joints are supplied complete with a stopper.
Ordering information: For replacement stoppers see CG-3000.