Flasks, Heavy Wall, Round Bottom, Single Neck, 2 to 20 L, Chemglass
Supplier: Chemglass
CG-1506-28 CG-1506-29 CG-1506-30 CG-1506-35 CG-1506-39 CG-1506-41 CG-1506-42 CG-1506-43
80068-796EA 542.75 CAD
80068-796 80068-798 80068-800 80068-808 80068-816 80068-820 80068-822 80068-824
Flasks, Heavy Wall, Round Bottom, Single Neck, 2 to 20 L, Chemglass
Flasks Round Bottom Flasks
Round bottom, heavy wall single short neck flask with a standard taper outer joint.
  • High-quality heavy wall flasks are short neck
  • Flasks feature a groove to aid in clamping of the flask
The 29/26 flasks have a groove tooled just below the grind to aid in clamping of the flask.