Flasks, Heavy Wall, Round Bottom, Threaded Inlet, Chemglass
Supplier: Chemglass
CG-1514-01 CG-1514-07 CG-1514-09 CG-1514-11 CG-1514-20 CG-1514-21 CG-1514-22 CG-1514-24
80069-056EA 289.53 CAD
80069-056 80069-058 80069-060 80069-062 80069-066 80069-068 80069-070 80069-072
Flasks, Heavy Wall, Round Bottom, Threaded Inlet, Chemglass
Flasks Round Bottom Flasks
Heavy wall flask with a standard taper outer joint and a number seven.
  • Heavy wall flasks feature side neck for insertion
  • Side neck insertion for thermometers or bleed tubes
Chem-Thread has a side neck for insertion of thermometers or bleed tubes having an O.D. between 4 and 7 mm.