Flasks, Recovery, Heavy Wall, Wide Mouth, Single Neck, GL 80, Chemglass
Supplier: Chemglass
CG-1512-X-02 CG-1512-X-13 CG-1512-X-12 CG-1512-X-03 CG-1512-X-01 CG-1512-X-11
10170-274EA 896.34 CAD
10170-274 10170-292 10170-636 10170-658 10171-126 10171-128
Flasks, Recovery, Heavy Wall, Wide Mouth, Single Neck, GL 80, Chemglass
Flasks Rotary Evaporator Flasks
Heavy wall, wide mouth flask with a GL 80 glass thread and a 24/40 connecting adapter.
  • Flasks have a heavy wall for vacuum applications and are balanced to prevent run-out during rotation
  • Unique design makes it easier to handle highly viscous liquids, powders, and solids
  • The large opening permits trouble-free emptying and cleaning and easy access of products with a spatula or policeman
The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation. Complete code supplied complete with PTFE GL 80 to 24/40 adapter. PTFE adapter has FDA grade silicone gasket behind a PTFE envelope type seal so that liquid or powders are only exposed to glass or PTFE.