Flasks, Heavy Wall, Apollo, Single Neck, Chemglass
Supplier: Chemglass
CG-1512-A-06 CG-1512-A-01 CG-1512-A-02 CG-1512-A-03 CG-1512-A-04 CG-1512-A-05
33502-618EA 178.51 CAD
33502-618 89028-582 89028-584 89028-586 89028-588 89028-590
Flasks, Heavy Wall, Apollo, Single Neck, Chemglass
Flasks Flat Bottom Flasks
Heavy wall single neck flask with a standard taper outer joint.
  • High-quality heavy wall flasks are short neck
  • Flasks feature a low profile design
  • Which allows the flask to sit upright on the bench
  • Permits easy access of products with a spatula or policeman
The flask contour is excellent for controlled evaporation of foaming solvents during rotary evaporation.