Condensers, Allihn, Chemglass
Supplier: Chemglass
CG-1207-03 CG-1207-14
80067-220EA 402.89 CAD
80067-220 80067-226
Condensers, Allihn, Chemglass
Condensers
Condensers are for use with CG-1368 and Allihn condensers with soxhlet extraction apparatus.
  • Allihn style reflux condenser
  • With a plain tubulation at the top
  • Lower inner drip tip joint
Hose connections have an O.D. of 10 mm at the middle serration.