Reaction Vessels, Cylindrical, Unjacketed, Large Scale, Chemglass
Supplier: Chemglass
CG-1924-30 CG-1924-31 CG-1924-32 CG-1924-33 CG-1924-34 CG-1924-35
16002-822EA 4635.28 CAD
16002-822 16002-824 16002-826 16002-828 16002-830 16002-832
Reaction Vessels, Cylindrical, Unjacketed, Large Scale, Chemglass
Reaction Vessels Reaction Vessels, Cylindrical Shape
Unjacketed large scale process reaction vessels.
  • These flush bottom (zero dead space) valves are designed to prevent solids from collecting in the bottom of the vessel
  • Vessels equipped with the 0-20 mm sealed on valve are supplied complete with a TFE plug having Perfluoro chemically resistant O-rings
  • Vessels are serial numbered and have a graduation scale on both sides showing the approximate volume
The 10, 15, and 20 l vessels have a 200 mm ID flange with an O-ring groove formed in the surface of the flange. The 30 l and 50 l have a 300 mm ID flatMore Product Information