Flasks, Tapered, European Style, 4-Necks, Threaded Inlet, Chemglass
Supplier: Chemglass
CG-1572-02 CG-1572-05 CG-1572-01 CG-1572-03
80070-328EA 523.09 CAD
80070-328 80070-334 80070-326 80070-330
Flasks, Tapered, European Style, 4-Necks, Threaded Inlet, Chemglass
Flasks Round Bottom Flasks
Flasks feature a tapered european design with 4-necks.
  • European style flask with tapered walls
  • A shallow hemispherical bottom for more efficient mechanical or magnetic stirring
With standard taper outer joints on center and side necks and a #7 Chem-Thread for insertion of thermometers or bleed tubes having an O.D. between 4 and 7 mm.