Digital Temperature Controllers, Controller Only, Chemglass

Supplier: Chemglass
CG-15001-11 CG-15001-12
89028-500EA 2774.86 CAD
89028-500 89028-502
Digital Temperature Controllers, Controller Only, Chemglass
Controllers Temperature Controllers
Temperature controller for applications that do not require the precise temperature (±0.1°).

  • Auto tune PID process controller
  • For use with Chemglass oil baths or mantles up to 5 l

Economical controller has 1500 watts of power and will control temperatures to ±1.0 °C.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR