Distilling Heads, Solvent, Modified, Chemglass
Supplier: Chemglass
CG-1233-03 CG-1233-05 CG-1233-01 CG-1233-02
80067-552EA 1583.02 CAD
80067-552 80067-556 80067-548 80067-550
Distilling Heads, Solvent, Modified, Chemglass
Distillation Components Distillation Heads
Designed to maintain distilled solvents in an inert atmosphere.
  • Top of main vapor tube is open
  • Additional vapor tube has been added to decrease risk of pressure buildup
  • Upper sampling port has a 4 mm PTFE stopcock with a 14/20 outer joint
  • For use with CG-3022 septa
Lower drain is centered to allow total drainage of storage head. Double oblique 2 mm bore PTFE stopcock on the 250 through 1000 ml sizes allows for removal of distilled solvent or return to flask. The 250 ml size has a 4 mm double oblique stopcock. Both the uptake tube and top outerMore Product Information