Condensers, Reflux, Coil Type, Chemglass
Supplier: Chemglass
CG-1213-L-20 CG-1213-L-21
80067-300EA 787.35 CAD
80067-300 80067-302
Condensers, Reflux, Coil Type, Chemglass
Condensers
Condensers feature a top outer standard taper joint.
  • Jacket has an O.D. of 41 mm while the coil O.D. is 8 mm
  • Excellent for use with high vapor pressure solvents
Similar to CG-1213, but having a larger diameter jacket and coil, thereby providing maximum cooling capacity, while being shorter in length. Condenser has a top outer standard taper joint and lower inner drip tip joint.